2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

052244-500MG 052244-2.5G
101898-862EA 364.62 USD
101898-862 101898-864
2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
2-Amino-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562017
CAS Number: 1236265-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR