2-Amino-N-benzyl-N-ethylpropanamide hydrochloride

Supplier: Matrix Scientific

052243-500MG 052243-2.5G
101898-858EA 364.62 USD
101898-858 101898-860
2-Amino-N-benzyl-N-ethylpropanamide hydrochloride
2-Amino-N-benzyl-N-ethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD13562016
CAS Number: 1246172-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR