2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-propanone hydrochloride

Supplier: Matrix Scientific

052239-500MG 052239-2.5G
101898-842EA 364.62 USD
101898-842 101898-844
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-propanone hydrochloride
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-propanone hydrochloride

Hazard: Irritant

Formula: C₉H₂₀ClN₃O₂
MDL Number: MFCD13562012
CAS Number: 1236258-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR