2-Amino-N-cyclohexyl-N-ethylpropanamide hydrochloride

Supplier: Matrix Scientific

052238-500MG 052238-2.5G
101898-838EA 364.62 USD
101898-838 101898-840
2-Amino-N-cyclohexyl-N-ethylpropanamide hydrochloride
2-Amino-N-cyclohexyl-N-ethylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562011
CAS Number: 1246172-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR