2-Amino-N-benzyl-N-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052237-500MG 052237-2.5G
101898-834EA 364.62 USD
101898-834 101898-836
2-Amino-N-benzyl-N-methylpropanamide hydrochloride
2-Amino-N-benzyl-N-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD13562010
CAS Number: 1236262-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR