2-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052236-500MG 052236-2.5G
101898-830EA 364.62 USD
101898-830 101898-832
2-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride
2-Amino-1-(2-ethyl-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562009
CAS Number: 1236260-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR