2-Amino-1-(4-hydroxy-1-piperidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052233-500MG 052233-2.5G
101898-818EA 364.62 USD
101898-818 101898-820
2-Amino-1-(4-hydroxy-1-piperidinyl)-1-propanone hydrochloride
2-Amino-1-(4-hydroxy-1-piperidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD13562006
CAS Number: 1236262-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR