4-(2-Aminopropanoyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(2-Aminopropanoyl)piperazin-2-one hydrochloride

052232-500MG 052232-2.5G
101898-814EA 364.62 USD
101898-814 101898-816
4-(2-Aminopropanoyl)-2-piperazinone hydrochloride
4-(2-Aminopropanoyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃O₂
MW: 207.66 g/mol
MDL Number: MFCD13562005
CAS Number: 1236262-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR