2-Amino-1-(1-azepanyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052231-500MG 052231-2.5G
101898-810EA 364.62 USD
101898-810 101898-812
2-Amino-1-(1-azepanyl)-1-propanone hydrochloride
2-Amino-1-(1-azepanyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562004
CAS Number: 1236260-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR