2-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052230-500MG 052230-2.5G
101898-806EA 364.62 USD
101898-806 101898-808
2-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride
2-Amino-1-(4-benzyl-1-piperazinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClN₃O
MDL Number: MFCD13562003
CAS Number: 1236265-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR