N,N-Diallyl-2-aminopropanamide hydrochloride

Supplier: Matrix Scientific

052225-500MG 052225-2.5G
101898-786EA 364.62 USD
101898-786 101898-788
N,N-Diallyl-2-aminopropanamide hydrochloride
N,N-Diallyl-2-aminopropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD13561998
CAS Number: 1236266-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR