2-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride
Supplier: Matrix Scientific
052224-500MG
052224-2.5G
101898-782EA
364.62
USD
101898-782
101898-784
2-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride
2-Amino-N-ethyl-N-(2-hydroxyethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13561997 CAS Number: 1236259-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...