2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride

Supplier: Matrix Scientific

052222-500MG 052222-2.5G
101898-774EA 364.62 USD
101898-774 101898-776
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-propanone hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O₂
MW: 194.66 g/mol
MDL Number: MFCD13561995
CAS Number: 1236260-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR