2-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052221-500MG
052221-2.5G
101898-770EA
364.62
USD
101898-770
101898-772
2-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
2-Amino-1-(4-morpholinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClN₂O₂ |
MDL Number:
MFCD13561994 CAS Number: 24152-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...