2-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride
Supplier: Matrix Scientific
052217-500MG
052217-2.5G
101898-754EA
364.62
USD
101898-754
101898-756
2-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride
2-Amino-1-(1-pyrrolidinyl)-1-propanone hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClN₂O |
MDL Number:
MFCD13561992 CAS Number: 24152-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...