2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-ethanone hydrochloride

Supplier: Matrix Scientific

052214-500MG 052214-2.5G
101898-742EA 364.62 USD
101898-742 101898-744
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-ethanone hydrochloride
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD13196038
CAS Number: 1210489-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR