2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052213-500MG 052213-2.5G
101898-738EA 384.62 USD
101898-738 101898-740
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-1-ethanone hydrochloride
2-Amino-1-(2,3-dihydro-1H-indol-1-yl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD13561990
CAS Number: 1220036-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR