2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-ethanone hydrochloride

Supplier: Matrix Scientific

052211-500MG 052211-2.5G
101898-730EA 364.62 USD
101898-730 101898-732
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-ethanone hydrochloride
2-Amino-1-[4-(2-hydroxyethyl)-1-piperazinyl]-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClN₃O₂
MDL Number: MFCD06411637
CAS Number: 1220017-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR