2-Amino-1-(2-ethyl-1-piperidinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052208-500MG 052208-2.5G
101898-718EA 364.62 USD
101898-718 101898-720
2-Amino-1-(2-ethyl-1-piperidinyl)-1-ethanone hydrochloride
2-Amino-1-(2-ethyl-1-piperidinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13561986
CAS Number: 1220018-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR