2-Amino-N-cyclohexyl-N-methylacetamide hydrochloride
Supplier: Matrix Scientific
052207-500MG
052207-2.5G
101898-714EA
364.62
USD
101898-714
101898-716
2-Amino-N-cyclohexyl-N-methylacetamide hydrochloride
2-Amino-N-cyclohexyl-N-methylacetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O |
MDL Number:
MFCD13561985 CAS Number: 1220036-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...