2-Amino-1-(4-benzyl-1-piperazinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052202-500MG 052202-2.5G
101898-694EA 364.62 USD
101898-694 101898-696
2-Amino-1-(4-benzyl-1-piperazinyl)-1-ethanone hydrochloride
2-Amino-1-(4-benzyl-1-piperazinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClN₃O
MDL Number: MFCD13561980
CAS Number: 1220018-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR