2-Amino-1-(3-methyl-1-piperidinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052199-500MG 052199-2.5G
101898-682EA 364.62 USD
101898-682 101898-684
2-Amino-1-(3-methyl-1-piperidinyl)-1-ethanone hydrochloride
2-Amino-1-(3-methyl-1-piperidinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13561978
CAS Number: 1220017-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR