2-Amino-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride
Supplier: Matrix Scientific
052198-500MG
052198-2.5G
101898-678EA
484.62
USD
101898-678
101898-680
2-Amino-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride
2-Amino-1-(2-methyl-1-piperidinyl)-1-ethanone hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O |
MDL Number:
MFCD13561977 CAS Number: 1220036-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...