2-Amino-N-ethyl-N-(2-hydroxyethyl)acetamide hydrochloride

Supplier: Matrix Scientific

052196-500MG 052196-2.5G
101898-670EA 364.62 USD
101898-670 101898-672
2-Amino-N-ethyl-N-(2-hydroxyethyl)acetamide hydrochloride
2-Amino-N-ethyl-N-(2-hydroxyethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13561975
CAS Number: 1220017-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR