2-Amino-N-butyl-N-methylacetamide hydrochloride

Supplier: Matrix Scientific

052195-500MG 052195-2.5G
101898-666EA 364.62 USD
101898-666 101898-668
2-Amino-N-butyl-N-methylacetamide hydrochloride
2-Amino-N-butyl-N-methylacetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13561974
CAS Number: 172596-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR