2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-ethanone hydrochloride

Supplier: Matrix Scientific

052194-500MG 052194-2.5G
101898-662EA 364.62 USD
101898-662 101898-664
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-ethanone hydrochloride
2-Amino-1-(3-hydroxy-1-pyrrolidinyl)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O₂
MDL Number: MFCD13561973
CAS Number: 1220017-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR