2-Amino-N-(2-hydroxyethyl)-N-methylacetamide hydrochloride

Supplier: Matrix Scientific

052192-500MG 052192-2.5G
101898-654EA 364.62 USD
101898-654 101898-656
2-Amino-N-(2-hydroxyethyl)-N-methylacetamide hydrochloride
2-Amino-N-(2-hydroxyethyl)-N-methylacetamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD13561972
CAS Number: 1220031-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR