6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

052188-500MG 052188-2.5G
101898-642EA 364.62 USD
101898-642 101898-644
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
6-Chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD13561971
CAS Number: 1220017-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR