N-Benzyl-6-chloro-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

052187-500MG 052187-2.5G
101898-638EA 364.62 USD
101898-638 101898-640
N-Benzyl-6-chloro-N-ethyl-2-pyridinamine
N-Benzyl-6-chloro-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂
MDL Number: MFCD13561970
CAS Number: 1220038-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR