1-(6-Chloro-2-pyridinyl)-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

052186-500MG 052186-2.5G
101898-634EA 364.62 USD
101898-634 101898-636
1-(6-Chloro-2-pyridinyl)-1,2,3,4-tetrahydroquinoline
1-(6-Chloro-2-pyridinyl)-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂
MDL Number: MFCD13561969
CAS Number: 1220036-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR