1-(6-Chloro-2-pyridinyl)indoline

Supplier: Matrix Scientific

052185-500MG 052185-2.5G
101898-630EA 364.62 USD
101898-630 101898-632
1-(6-Chloro-2-pyridinyl)indoline
1-(6-Chloro-2-pyridinyl)indoline

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂
MDL Number: MFCD13561968
CAS Number: 1220036-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR