2-[4-(6-Chloro-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

Synonyms: 2-(4-(6-Chloropyridin-2-yl)piperazin-1-yl)ethanol

052183-500MG 052183-2.5G
101898-622EA 364.62 USD
101898-622 101898-624
2-[4-(6-Chloro-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(6-Chloro-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MW: 241.72 g/mol
MDL Number: MFCD13561966
CAS Number: 1220038-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR