6-Chloro-N-cyclohexyl-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

052182-500MG 052182-2.5G
101898-618EA 364.62 USD
101898-618 101898-620
6-Chloro-N-cyclohexyl-N-ethyl-2-pyridinamine
6-Chloro-N-cyclohexyl-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD13561965
CAS Number: 1220017-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR