N-Benzyl-6-chloro-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

052181-500MG 052181-2.5G
101898-614EA 364.62 USD
101898-614 101898-616
N-Benzyl-6-chloro-N-methyl-2-pyridinamine
N-Benzyl-6-chloro-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂
MDL Number: MFCD13561964
CAS Number: 1219972-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR