1-(6-Chloro-2-pyridinyl)-2-ethylpiperidine

Supplier: Matrix Scientific

052180-500MG 052180-2.5G
101898-610EA 364.62 USD
101898-610 101898-612
1-(6-Chloro-2-pyridinyl)-2-ethylpiperidine
1-(6-Chloro-2-pyridinyl)-2-ethylpiperidine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD13561963
CAS Number: 1219982-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR