6-Chloro-N-cyclohexyl-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

052179-500MG 052179-2.5G
101898-606EA 364.62 USD
101898-606 101898-608
6-Chloro-N-cyclohexyl-N-methyl-2-pyridinamine
6-Chloro-N-cyclohexyl-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD13561962
CAS Number: 1220036-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR