1-(6-Chloro-2-pyridinyl)-4-piperidinol

Supplier: Matrix Scientific

Synonyms: 1-(6-Chloropyridin-2-yl)piperidin-4-ol

052177-500MG 052177-2.5G
101898-598EA 384.62 USD
101898-598 101898-600
1-(6-Chloro-2-pyridinyl)-4-piperidinol
1-(6-Chloro-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD12923030
CAS Number: 1220036-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR