1-(6-Chloro-2-pyridinyl)azepane

Supplier: Matrix Scientific

052175-500MG 052175-2.5G
101898-590EA 364.62 USD
101898-590 101898-592
1-(6-Chloro-2-pyridinyl)azepane
1-(6-Chloro-2-pyridinyl)azepane

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂
MDL Number: MFCD13561959
CAS Number: 1220038-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR