1-(6-Chloro-2-pyridinyl)-4-methylpiperazine

Supplier: Matrix Scientific

Synonyms: 1-(6-Chloropyridin-2-yl)-4-methylpiperazine

052173-500MG 052173-2.5G
101898-582EA 364.62 USD
101898-582 101898-584
1-(6-Chloro-2-pyridinyl)-4-methylpiperazine
1-(6-Chloro-2-pyridinyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MW: 211.7 g/mol
MDL Number: MFCD00925758
CAS Number: 153976-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR