1-(6-Chloro-2-pyridinyl)-3-methylpiperidine

Supplier: Matrix Scientific

052171-500MG 052171-2.5G
101898-574EA 364.62 USD
101898-574 101898-576
1-(6-Chloro-2-pyridinyl)-3-methylpiperidine
1-(6-Chloro-2-pyridinyl)-3-methylpiperidine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂
MDL Number: MFCD13561957
CAS Number: 1220017-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR