1-(6-Chloro-2-pyridinyl)-2-methylpiperidine

Supplier: Matrix Scientific

052170-500MG 052170-2.5G
101898-570EA 364.62 USD
101898-570 101898-572
1-(6-Chloro-2-pyridinyl)-2-methylpiperidine
1-(6-Chloro-2-pyridinyl)-2-methylpiperidine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂
MDL Number: MFCD13561956
CAS Number: 1220030-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR