N,N-Diallyl-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

052169-500MG 052169-2.5G
101898-566EA 364.62 USD
101898-566 101898-568
N,N-Diallyl-6-chloro-2-pyridinamine
N,N-Diallyl-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂
MDL Number: MFCD13561955
CAS Number: 1220031-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR