2-[(6-Chloro-2-pyridinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052168-500MG 052168-2.5G
101898-562EA 364.62 USD
101898-562 101898-564
2-[(6-Chloro-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(6-Chloro-2-pyridinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD13561954
CAS Number: 1219982-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR