N-Butyl-6-chloro-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

052167-500MG 052167-2.5G
101898-558EA 364.62 USD
101898-558 101898-560
N-Butyl-6-chloro-N-methyl-2-pyridinamine
N-Butyl-6-chloro-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂
MDL Number: MFCD13561953
CAS Number: 1220017-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR