1-(6-Chloro-2-pyridinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

052166-500MG 052166-2.5G
101898-554EA 461.54 USD
101898-554 101898-556
1-(6-Chloro-2-pyridinyl)-3-pyrrolidinol
1-(6-Chloro-2-pyridinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD13561952
CAS Number: 1219972-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR