4-(6-Chloro-2-pyridinyl)morpholine
Supplier: Matrix Scientific
052165-500MG
052165-2.5G
101898-550EA
584.62
USD
101898-550
101898-552
4-(6-Chloro-2-pyridinyl)morpholine
4-(6-Chloro-2-pyridinyl)morpholine
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O MW: 198.65 g/mol |
MDL Number:
MFCD11848425 CAS Number: 330682-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...