N-(6-Chloro-2-pyridinyl)-N,N-diethylamine

Supplier: Matrix Scientific

052162-500MG 052162-2.5G
101898-538EA 186.15 USD
101898-538 101898-540
N-(6-Chloro-2-pyridinyl)-N,N-diethylamine
N-(6-Chloro-2-pyridinyl)-N,N-diethylamine

Hazard: Irritant

Formula: C₉H₁₃ClN₂
MDL Number: MFCD13561949
CAS Number: 1220017-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR