2-Chloro-6-(1-pyrrolidinyl)pyridine

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-(pyrrolidin-1-yl)pyridine

052161-500MG 052161-2.5G
101898-534EA 364.62 USD
101898-534 101898-536
2-Chloro-6-(1-pyrrolidinyl)pyridine
2-Chloro-6-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
MDL Number: MFCD04625620
CAS Number: 117362-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR