2-[4-(5-Bromo-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

052154-500MG 052154-2.5G
101898-506EA 166.15 USD
101898-506 101898-508
2-[4-(5-Bromo-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(5-Bromo-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₆BrN₃O
MDL Number: MFCD03426067
CAS Number: 364794-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR