2-(N-Benzyl-N-methylamino)-5-bromopyridine

Supplier: Matrix Scientific

052152-500MG 052152-2.5G
101898-498EA 364.62 USD
101898-498 101898-500
2-(N-Benzyl-N-methylamino)-5-bromopyridine
2-(N-Benzyl-N-methylamino)-5-bromopyridine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂
MW: 263.14 g/mol
MDL Number: MFCD12913988
CAS Number: 1187386-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR